missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGTA Mouse anti-Human, Mouse, Clone: 3B10B11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60305-1-IG-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein which is capable of interacting with the major nonstructural protein of parvovirus H-1 and 70-kDa heat shock cognate protein; however, its function is not known. Since this transcript is expressed ubiquitously in various tissues, this protein may serve a housekeeping function.Specifications
| SGTA | |
| Monoclonal | |
| 2.03 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43765, Q8BJU0 | |
| Sgta | |
| SGTA Fusion Protein Ag1475 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 3B10B11 | |
| Unconjugated | |
| SGTA | |
| Alpha SGT, alphaSGT, hSGT, SGT, SGT1, SGTA, UBP, Vpu binding protein | |
| Mouse | |
| Protein A | |
| RUO | |
| 52551, 6449 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction