missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGTA Mouse anti-Human, Mouse, Clone: 3B10B11, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | SGTA |
|---|---|
| Clone | 3B10B11 |
| Concentration | 2.03 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16807395
|
Proteintech
60305-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16897385
|
Proteintech
60305-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a protein which is capable of interacting with the major nonstructural protein of parvovirus H-1 and 70-kDa heat shock cognate protein; however, its function is not known. Since this transcript is expressed ubiquitously in various tissues, this protein may serve a housekeeping function.Specifications
| SGTA | |
| 2.03 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Alpha SGT, alphaSGT, hSGT, SGT, SGT1, SGTA, UBP, Vpu binding protein | |
| Sgta | |
| IgG2b | |
| Protein A | |
| Antibody |
| 3B10B11 | |
| Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Mouse, Human | |
| O43765, Q8BJU0 | |
| 52551, 6449 | |
| SGTA Fusion Protein Ag1475 | |
| Primary | |
| -20°C | |
| SGTA |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title