Learn More
Trajan™ CTC PAL HPLC Fixed Needle Autosampler Syringe
Borosilicate glass barrel and stainless steel needles provide strength and inertness. Backing provides quick and accurate volume assessment.
Brand:  Trajan™ 005720
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 12967526
  • 1225.00 NOK / Each
Estimated Shipment: 01-07-2024
Description
  • High temperature syringes for headspace application with temperatures up to 150°C
  • PTFE plunger seal option for smooth, leak free operation - suitable for both liquid and gas samples, with replaceable plungers
  • Specifications of the syringes are exactly matched to CTC holders to ensure proper fit and reliable functioning
Specifications
Specifications
22 G
Fixed Needle Syringe
100 μL
LC
0.72 mm
100F-CTC-GT-LC(0.4)
51 mm