Learn More
Trajan™ CTC PAL HPLC Fixed Needle Autosampler Syringe
Borosilicate glass barrel and stainless steel needles provide strength and inertness. Backing provides quick and accurate volume assessment.
1225.00 NOK - 1465.00 NOK
Product Code | Brand | Volume (Metric) | Length (Metric) Needle | Model | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Length (Metric) Needle | Model | Price | Quantity & Availability | |||
12947556
|
Trajan™
005715 |
100 μL | 51 mm | 100F-CTC-GT-LC |
1225.00 NOK
Each |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12967526
|
Trajan™
005720 |
100 μL | 51 mm | 100F-CTC-GT-LC(0.4) |
1225.00 NOK
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||
12987526
|
Trajan™
007720 |
500 μL | 51 mm | 500F-CTC-GT-LC(0.4) |
1250.00 NOK
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||
12957526
|
Trajan™
003715 |
25 μL | 51 mm | 25F-CTC-GT-LC |
1465.00 NOK
Each |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- High temperature syringes for headspace application with temperatures up to 150°C
- PTFE plunger seal option for smooth, leak free operation - suitable for both liquid and gas samples, with replaceable plungers
- Specifications of the syringes are exactly matched to CTC holders to ensure proper fit and reliable functioning
Specifications
22 G | |
51 mm |