Learn More
Novus Biologicals™ SF-1/NR5A1/Steroidogenic Factor 1 Overexpression Lysate
Shop All Bio Techne ProductsDescription
The SF-1/NR5A1/Steroidogenic Factor 1 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
| Cell Line | The lysate was created in HEK293T cells, using plasmid ID RC27577 and based on accession number NM_4959. The protein contains a C-terminal DDK tag. |
| For Use With (Application) | Western Blot |
| Quantity | 0.1 mg |
| Storage Buffer | RIPA buffer |
| Purity | Protein |
| Test Specificity | Homo sapiens nuclear receptor subfamily 5, group A, member 1 (NR5A1), mRNA. |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Protein State | Native |
For Research Use Only