Learn More
SERPINB9 Mouse anti-Human, Unconjugated, Clone: 2H9D2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67950-1-IG-150UL
 View more versions of this product
Product Code. 17551456
  • 4836.10 NOK / 150µL
Description
The SERPINB9 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human SERPINB9. This antibody recognizes human, pig antigen. The SERPINB9 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
SERPINB9
Monoclonal
Unconjugated
150 μL
5272
IgG1
Western Blot, Immunohistochemistry
2H9D2
Mouse
Primary
Human, Pig