Learn More
SERPINB9 Mouse anti-Human, Unconjugated, Clone: 2H9D2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67950-1-IG-150UL
This item is not returnable.
View return policy
Description
The SERPINB9 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human SERPINB9. This antibody recognizes human, pig antigen. The SERPINB9 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| SERPINB9 | |
| Monoclonal | |
| Unconjugated | |
| 150 μL | |
| 5272 | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| 2H9D2 | |
| Mouse | |
| Primary | |
| Human, Pig |