missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINB9 Mouse anti-Human, Unconjugated, Clone: 2H9D2, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | SERPINB9 |
|---|---|
| Clone | 2H9D2 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17561456
|
Proteintech
67950-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17551456
|
Proteintech
67950-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SERPINB9 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human SERPINB9. This antibody recognizes human, pig antigen. The SERPINB9 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| SERPINB9 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Human, Pig | |
| IgG1 |
| 2H9D2 | |
| Monoclonal | |
| Mouse | |
| 5272 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title