missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ SERPINB6 (Human) Recombinant Protein
Human SERPINB6 full-length ORF ( AAH01394, 1 a.a. - 376 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005269-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MDVLAEANGTFALNLLKTLGKDNSKNVFFSPMSMSCALAMVYMGAKGNTAAQMAQILSFNKSGGGGDIHQGFQSLLTEVNKTGTQYLLRVANRLFGEKSCDFLSSFRDSCQKFYQAEMEELDFISAVEKSRKHINTWVAEKTEGKIAELLSPGSVDPLTRLVLVNAVYFRGNWDEQFDKENTEERLFKVSKNEEKPVQMMFKQSTFKKTYIGEIFTQILVLPYVGKELNMIIMLPDETTDLRTVEKELTYEKFVEWTRLDMMDEEEVEVSLPRFKLEESYDMESVLRNLGMTDAFELGKADFSGMSQTDLSLSKVVHKSFVEVNEEGTEAAAATAAIMMMRCARFVPRFCADHPFLFFIQHSKTNGILFCGRFSSP
Specifications
AAH01394 | |
serpin peptidase inhibitor, clade B (ovalbumin), member 6 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SERPINB6 | |
GST |
5269 | |
Wheat germ expression system | |
10 μg | |
CAP, DKFZp686I04222, MGC111370, MSTP057, PI6, PTI, SPI3 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |