missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ SERPINB6 (Human) Recombinant Protein
Human SERPINB6 full-length ORF ( AAH01394, 1 a.a. - 376 a.a.) recombinant protein with GST-tag at N-terminal.
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAH01394 |
---|---|
Gene ID (Entrez) | 5269 |
Name | serpin peptidase inhibitor, clade B (ovalbumin), member 6 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16189061
|
Abnova™
H00005269-P01.10UG |
10 μg |
4115.00 NOK
10µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16199061
|
Abnova™
H00005269-P01.25UG |
25 μg |
6245.00 NOK
25µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: MDVLAEANGTFALNLLKTLGKDNSKNVFFSPMSMSCALAMVYMGAKGNTAAQMAQILSFNKSGGGGDIHQGFQSLLTEVNKTGTQYLLRVANRLFGEKSCDFLSSFRDSCQKFYQAEMEELDFISAVEKSRKHINTWVAEKTEGKIAELLSPGSVDPLTRLVLVNAVYFRGNWDEQFDKENTEERLFKVSKNEEKPVQMMFKQSTFKKTYIGEIFTQILVLPYVGKELNMIIMLPDETTDLRTVEKELTYEKFVEWTRLDMMDEEEVEVSLPRFKLEESYDMESVLRNLGMTDAFELGKADFSGMSQTDLSLSKVVHKSFVEVNEEGTEAAAATAAIMMMRCARFVPRFCADHPFLFFIQHSKTNGILFCGRFSSP
Specifications
AAH01394 | |
serpin peptidase inhibitor, clade B (ovalbumin), member 6 | |
125% SDS-PAGE Stained with Coomassie Blue | |
CAP, DKFZp686I04222, MGC111370, MSTP057, PI6, PTI, SPI3 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
5269 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SERPINB6 | |
GST |