missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERINC5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27066-1-AP-150UL
Additional Details : Weight : 0.00010kg
Specifications
SERINC5 | |
Polyclonal | |
Unconjugated | |
SERINC5 | |
C5orf12, SERINC5, serine incorporator 5, TPO1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
256987 | |
-20°C | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q86VE9 | |
SERINC5 | |
SERINC5 Fusion Protein Ag25265 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |