missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERINC5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | SERINC5 |
---|---|
Concentration | 0.4 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16885355
|
Proteintech
27066-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16875355
|
Proteintech
27066-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SERINC5 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q86VE9 | |
256987 | |
SERINC5 Fusion Protein Ag25265 | |
Primary | |
-20°C | |
SERINC5 |
0.4 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C5orf12, SERINC5, serine incorporator 5, TPO1 | |
SERINC5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |