Learn More
SERINC5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27066-1-AP-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16885355
  • 1611.84 NOK / 20µL
Estimated Shipment: 13-08-2024
Specifications
SERINC5
Polyclonal
Unconjugated
SERINC5
C5orf12, SERINC5, serine incorporator 5, TPO1
Rabbit
Antigen Affinity Chromatography
RUO
256987
-20°C
Liquid
Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q86VE9
SERINC5
SERINC5 Fusion Protein Ag25265