Specifications
| Quantity | 100 g |
| Product Type | Sephadex G50 Media |
| Format | Solid Bead |
| For Use With (Application) | Desalting and Buffer exchange of biomolecules |
| Support Type | Cross-linked Dextron |
| Sample Type | DNA |
| Content And Storage | 4° to 30°C |