Learn More
WTW™ SenTix™ SP-DIN PLUS Special pH Electrode
Click to view available options
Material:
Plastic Shaft
Unit Size:
Each
Specifications
Specifications
| Connector Types | Plug Head |
| Material | Plastic Shaft |
| pH Range | 2 to 13 |
| Temperature (Metric) Operating | 0°C to 80°C |
| Type | SenTix™ PLUS Special pH Electrode |
| For Use With (Equipment) | pH meter |
| Includes | DIN electrode plug |
| Model | SenTix™ SP-DIN |
| Temperature (Metric) Range | 0°C to 80°C |
| Length (Metric) Stem | 65/25 mm |