Learn More
WTW™ SenTix™ 21 pH Electrode
Product Code. 10635241
Change view
Click to view available options
Connector Types:
DIN
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Unit Size
10635241 DIN Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10635241 Supplier WTW™ Supplier No. 103631
  • ONLINE EXCLUSIVE PRICE
    2000.00 NOK
    1888.41 NOK / Pack of 1
    Save 111.59 NOK
    5% Off
View all promotions
Estimated Shipment: 30-11-2026
Recommended for all applications where reliable measured values are needed even under the most adverse conditions. For demanding measurements in the lab, there are SenTix™ electrodes with 3 mol/l KCl liquid electrolyte, easy to clean glass shaft and platinum diaphragm.
  • Ideal for portable measurement but also for routine measurement in the lab 
  • SenTix™: Standard electrodes with gel and precision electrodes with liquid gel electrolyte for pH measurements in lab applications and in the field
  • Maintenance-free gel reference system across the entire product life
  • Quick response time by good connection to the medium for electrodes with liquid filled reference system
  • For demanding measurements in the lab, there are SenTix™ electrodes with 3 mol/l KCl liquid electrolyte, easy to clean glass shaft and platinum diaphragm
TRUSTED_SUSTAINABILITY
Specifications
Connector Types DIN
Junction Fiber
Material Plastic Shaft, Fiber Diaphragm
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 80°C
Type SenTix™ pH Electrode
For Use With (Equipment) Vario pH meter
Includes DIN electrode plug, 1m cable
Model SenTix™ 21
Storage Requirements Immerse the combination electrode in reference electrolyte (KCl 3mol/L, Ag+ free), pH combination electrodes must not be stored dry or in deionized water
Temperature (Metric) Range 0°C to 80°C
Length (Metric) Cable 1 m
Length (Metric) Stem 120 mm