Learn More
Secnidazole, TRC
Product Code. 17922525
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17922525 100 mg 100mg
17944915 10 mg 10mg
17922055 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17922525 Supplier Toronto Research Chemicals Supplier No. TRCS225000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3366-95-8
Molecular Formula C7 H11 N3 O3
Molecular Weight (g/mol) 185.18
Synonym Secnidazole, 1H-Imidazole-1-ethanol, α,2-dimethyl-5-nitro-, Imidazole-1-ethanol, α,2-dimethyl-5-nitro- (7CI,8CI), α,2-Dimethyl-5-nitro-1H-imidazole-1-ethanol, 1-(2-Methyl-5-nitro-1H-imidazol-1-yl)propan-2-ol, 1-(2-Methyl-5-nitroimidazol-1-yl)-2-propanol, Deprozol, Flagentyl, PM 185184, RP 14539, SYM 1219, Sabima, Secnidazole, Secnol, Solosec
IUPAC Name 1-(2-methyl-5-nitroimidazol-1-yl)propan-2-ol
SMILES CC(O)Cn1c(C)ncc1[N+](=O)[O-]
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C7H11N3O3/c1-5(11)4-9-6(2)8-3-7(9)10(12)13/h3,5,11H,4H2,1-2H3
Formula Weight 185.08
Percent Purity >95%
Chemical Name or Material Secnidazole
Grade HPLC