Learn More
SEC5/EXOC2 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1C11G2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66011-1-IG-150UL
 View more versions of this product
Product Code. 16847505
  • 4836.10 NOK / 150µL
Description
The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and the functions of the exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. This interaction has been shown to mediate filopodia formation in fibroblasts.
Specifications
Specifications
SEC5/EXOC2
Monoclonal
1.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O54921, Q96KP1, Q9D4H1
Exoc2
SEC5/EXOC2 Fusion Protein Ag17866
150 μL
Primary
Human, Mouse, Pig, Rat
Antibody
IgG2b
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
1C11G2
Unconjugated
EXOC2
EXOC2, exocyst complex component 2, Exocyst complex component Sec5, SEC5, SEC5L1, Sec5p
Mouse
Protein A
RUO
100520314, 171455, 55770, 66482
-20°C