missing translation for 'onlineSavingsMsg'
Learn More

SEC5/EXOC2 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1C11G2, Proteintech

Product Code. p-7212701 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16857505 20 μL 20µL
16847505 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16857505

Brand: Proteintech 660111IG20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and the functions of the exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. This interaction has been shown to mediate filopodia formation in fibroblasts.

Specifications

Antigen SEC5/EXOC2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1C11G2
Concentration 1.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EXOC2
Gene Accession No. O54921, Q96KP1, Q9D4H1
Gene Alias EXOC2, exocyst complex component 2, Exocyst complex component Sec5, SEC5, SEC5L1, Sec5p
Gene Symbols Exoc2
Host Species Mouse
Immunogen SEC5/EXOC2 Fusion Protein Ag17866
Purification Method Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100520314, 171455, 55770, 66482
Target Species Rat, Mouse, Human, Pig
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.