Learn More
Kern Sohn School Balance EMS
Entry level laboratory balance with large weighing plate.
Brand:  Kern Sohn EMS 300-3
Additional Details : Weight : 1.40000kg
 View more versions of this product
Product Code. 17351603
  • 3483.94 NOK / Each
Estimated Shipment: 07-05-2024
Description
  • Especially suitable for use in schools and universities, for example for biology, chemistry, physics
  • Large, shock proof weighing plate made of plastic
  • Particularly flat design
  • Ergonomically optimised key pad with large keys and a high-contrast LCD display
  • Secure and non-slip positioning through rubber feet
  • Adjusting program CAL for quick setting of the balance accuracy, external test weights at an additional price, see Test weights
Specifications
Specifications
300 g
0.005 g
School Balance
105 mm
0.001 g
0.002 g
105 mm
Universal Mains Adapter