Learn More
Kern Sohn School Balance EMS
Entry level laboratory balance with large weighing plate.
1142.28 NOK - 3483.94 NOK
Specifications
Type School Balance
Electrical Requirements Universal Mains Adapter
View More Specs
Products 6
Product Code Brand Capacity Readability Price Quantity & Availability  
Product Code Brand Capacity Readability Price Quantity & Availability  
17331603
View Documents Promotion Details
Kern Sohn
EMS 12K0.1
12 kg 0.1 g
2684.34 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-04-2024
to see stock.
17341603
View Documents Promotion Details
Kern Sohn
EMS 12K1
12 kg 1 g
1199.39 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-04-2024
to see stock.
16318147
View Documents Promotion Details
Kern Sohn
EMS 3000-2
3000 g 0.01 g
3027.03 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-04-2024
to see stock.
17351603
View Documents Promotion Details
Kern Sohn
EMS 300-3
300 g 0.001 g
3483.94 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-04-2024
to see stock.
17361603
View Documents Promotion Details
Kern Sohn
EMS 6K1
6000 g 1 g
1142.28 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-04-2024
to see stock.
16456440
View Documents Promotion Details
Kern Sohn
EMS6K0.1
6000 g 0.1 g
2170.33 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-04-2024
Description
  • Especially suitable for use in schools and universities, for example for biology, chemistry, physics
  • Large, shock proof weighing plate made of plastic
  • Particularly flat design
  • Ergonomically optimised key pad with large keys and a high-contrast LCD display
  • Secure and non-slip positioning through rubber feet
  • Adjusting program CAL for quick setting of the balance accuracy, external test weights at an additional price, see Test weights
Specifications
Specifications
School Balance
Universal Mains Adapter