Learn More
Kern Sohn School Balance EMS
Entry level laboratory balance with large weighing plate.
1142.28 NOK - 3483.94 NOK
Product Code | Brand | Capacity | Readability | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Readability | Price | Quantity & Availability | ||||
17331603
|
Kern Sohn
EMS 12K0.1 |
12 kg | 0.1 g |
2684.34 NOK
Each |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17341603
|
Kern Sohn
EMS 12K1 |
12 kg | 1 g |
1199.39 NOK
Each |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16318147
|
Kern Sohn
EMS 3000-2 |
3000 g | 0.01 g |
3027.03 NOK
Each |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17351603
|
Kern Sohn
EMS 300-3 |
300 g | 0.001 g |
3483.94 NOK
Each |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17361603
|
Kern Sohn
EMS 6K1 |
6000 g | 1 g |
1142.28 NOK
Each |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16456440
|
Kern Sohn
EMS6K0.1 |
6000 g | 0.1 g |
2170.33 NOK
Each |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Especially suitable for use in schools and universities, for example for biology, chemistry, physics
- Large, shock proof weighing plate made of plastic
- Particularly flat design
- Ergonomically optimised key pad with large keys and a high-contrast LCD display
- Secure and non-slip positioning through rubber feet
- Adjusting program CAL for quick setting of the balance accuracy, external test weights at an additional price, see Test weights
Specifications
School Balance | |
Universal Mains Adapter |