Learn More
Kern Sohn School Balance EMS
Entry level laboratory balance with large weighing plate.
Brand: Kern Sohn EMS6K0.1
Additional Details : Weight : 1.00000kg
Description
- Especially suitable for use in schools and universities, for example for biology, chemistry, physics
- Large, shock proof weighing plate made of plastic
- Particularly flat design
- Ergonomically optimised key pad with large keys and a high-contrast LCD display
- Secure and non-slip positioning through rubber feet
- Adjusting program CAL for quick setting of the balance accuracy, external test weights at an additional price, see Test weights
Specifications
6000 g | |
0.3 g | |
School Balance | |
190 mm |
0.1 g | |
0.1 g | |
175 mm | |
Universal Mains Adapter |