missing translation for 'onlineSavingsMsg'
Learn More

SCGB3A2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1710.33 NOK - 4836.10 NOK

Specifications

Antigen SCGB3A2
Concentration 600 μg/mL
Applications Immunohistochemistry, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16844745
View Documents
Proteintech
26228-1-AP-20UL
20 μL
1710.33 NOK
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16834745
View Documents
Proteintech
26228-1-AP-150UL
150 μL
4836.10 NOK
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The SCGB3A2 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human SCGB3A2. This antibody recognizes human antigen. The SCGB3A2 antibody has been validated for the following applications: IHC, ELISA analysis.

Specifications

Specifications

SCGB3A2
Immunohistochemistry, ELISA
Unconjugated
Human
IgG
600 μg/mL
Polyclonal
Rabbit
117156
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.