Learn More
SCGB3A2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26228-1-AP-20UL
This item is not returnable.
View return policy
Description
The SCGB3A2 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human SCGB3A2. This antibody recognizes human antigen. The SCGB3A2 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| SCGB3A2 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 117156 | |
| IgG |
| Immunohistochemistry, ELISA | |
| 600 μg/mL | |
| Rabbit | |
| Primary | |
| Human |