Learn More
SCGB3A2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26228-1-AP-20UL
 View more versions of this product
Product Code. 16844745
  • 1710.33 NOK / 20µL
Description
The SCGB3A2 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human SCGB3A2. This antibody recognizes human antigen. The SCGB3A2 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
SCGB3A2
Polyclonal
Unconjugated
20 μL
117156
IgG
Immunohistochemistry, ELISA
600 μg/mL