Learn More
Scandium(III) bromide, ultra dry, 99.99% (metals basis), Thermo Scientific™
2885.00 NOK - 9040.00 NOK
Chemical Identifiers
| CAS | 13465-59-3 |
|---|---|
| Molecular Formula | Br3Sc |
| Molecular Weight (g/mol) | 284.67 |
| MDL Number | MFCD00016324 |
| InChI Key | APPHYFNIXVIIJR-UHFFFAOYSA-K |
| Synonym | scandium tribromide, scandium bromide scbr3, scandium iii bromide, scandium iii bromide hydrate, scandium iii bromide, anhydrous, powder trace metals basis |
| PubChem CID | 83495 |
| IUPAC Name | scandium(3+) tribromide |
| SMILES | [Sc+3].[Br-].[Br-].[Br-] |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15477465
|
Thermo Scientific Alfa Aesar
047199.03 |
1g |
2885.00 NOK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15487465
|
Thermo Scientific Alfa Aesar
047199.06 |
5g |
9040.00 NOK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Ampouled under argon
Chemical Identifiers
| 13465-59-3 | |
| 284.67 | |
| APPHYFNIXVIIJR-UHFFFAOYSA-K | |
| 83495 | |
| [Sc+3].[Br-].[Br-].[Br-] |
| Br3Sc | |
| MFCD00016324 | |
| scandium tribromide, scandium bromide scbr3, scandium iii bromide, scandium iii bromide hydrate, scandium iii bromide, anhydrous, powder trace metals basis | |
| scandium(3+) tribromide |
Specifications
| 13465-59-3 | |
| MFCD00016324 | |
| APPHYFNIXVIIJR-UHFFFAOYSA-K | |
| scandium(3+) tribromide | |
| 83495 | |
| 99.99% | |
| Hygroscopic | |
| Scandium(III) bromide, Ultra dry |
| Br3Sc | |
| scandium tribromide, scandium bromide scbr3, scandium iii bromide, scandium iii bromide hydrate, scandium iii bromide, anhydrous, powder trace metals basis | |
| [Sc+3].[Br-].[Br-].[Br-] | |
| 284.67 | |
| 284.67 | |
| Ampule | |
| (metals basis) |
Safety and Handling
missing translation for 'einecsNumber' : 236-699-6
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Ampouled under argon
Recommended Storage : Ambient temperatures