CAS RN 13465-59-3
CAS RN 13465-59-3
IUPAC Name:
scandium(3+) tribromide
Synonyms:
scandium bromide
Molecular Weight (g/mol):
284.67
Molecular Formula:
Br3Sc
InChi Key:
APPHYFNIXVIIJR-UHFFFAOYSA-K
SMILES:
[Sc+3].[Br-].[Br-].[Br-]
1
–
1
of
1
results
1
Scandium(III) bromide, ultra dry, 99.99% (metals basis), Thermo Scientific™
CAS: 13465-59-3 Molecular Formula: Br3Sc Molecular Weight (g/mol): 284.67 MDL Number: MFCD00016324 InChI Key: APPHYFNIXVIIJR-UHFFFAOYSA-K Synonym: scandium tribromide,scandium bromide scbr3,scandium iii bromide,scandium iii bromide hydrate,scandium iii bromide, anhydrous, powder trace metals basis PubChem CID: 83495 IUPAC Name: scandium(3+) tribromide SMILES: [Sc+3].[Br-].[Br-].[Br-]