missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Spike Protein Monoclonal Antibody (OTI3H6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA814467
Additional Details : Weight : 0.05000kg
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
SARS-CoV-2 Spike Protein Monoclonal antibody specifically detects SARS-CoV-2 Spike Protein in Virus samples. It is validated for ELISA, Western BlotSpecifications
SARS-CoV-2 Spike Protein | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Mouse | |
Protein A/G | |
RUO | |
Virus | |
Antibody | |
IgG2b |
ELISA, Western Blot | |
OTI3H6 | |
Unconjugated | |
Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2 | |
Recombinant SARS-CoV-2 Spike-RBD protein produced in E.coli. | |
100 μL | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |