missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Spike Protein Monoclonal Antibody (OTI3H6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
4065.15 NOK
Specifications
Antigen | SARS-CoV-2 Spike Protein |
---|---|
Clone | OTI3H6 |
Concentration | 1 mg/mL |
Applications | ELISA, Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17549224
|
Origene Technologies
TA814467S |
30 μL |
4065.15 NOK
30µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17529224
|
Origene Technologies
TA814467 |
100 μL |
4065.15 NOK
100µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
SARS-CoV-2 Spike Protein Monoclonal antibody specifically detects SARS-CoV-2 Spike Protein in Virus samples. It is validated for ELISA, Western BlotSpecifications
SARS-CoV-2 Spike Protein | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Recombinant SARS-CoV-2 Spike-RBD protein produced in E.coli. | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles |
OTI3H6 | |
ELISA, Western Blot | |
Unconjugated | |
Mouse | |
Virus | |
Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2 | |
IgG2b | |
Protein A/G | |
Antibody |