Learn More
Gilson™ Pharmed™ Pump Tubing
Shop All Gilson Products
Click to view available options
Diameter (Metric) Inner:
0.5 mm
Unit Size:
Pack of 4
Description
- Polypropylene
Specifications
Specifications
| Diameter (Metric) Inner | 0.5 mm |
| Material | Santoprene |
| Overall Length (Metric) | 40 cm |
| For Use With | For MF1 and MF4 Pump Heads |
| Packaging Quantity | 4 Pack |
| Flow Rate | 2.5mL/min. |
| Product Type | Pump Tubing |