Learn More
Cole-Parmer™ Sanitary Coupling with Barb, Polysulfone
Cole-Parmer™ Sanitary Coupling with Barb, Polysulfone has a tapered funnel design to provide a steady fluid flow.
Brand:  Cole-Parmer™ 31512-07
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12469650
  • 1050.00 NOK / Each
Estimated Shipment: 13-02-2025
Description
  • Reduces dead space and improves flow.
  • Tapered funnel design provides a steady fluid flow.
  • Smooth inner surface prevents particle entrapment.
  • Seamless construction prevents contamination by eliminating leaks and areas of potential bacterial growth.
  • Material meets USP Class VI requirements and is FDA compliant for food contact.
Specifications
Specifications
Meets USP Class VI requirements and is FDA compliant for food contact
Polysulfone
FDA compliant for food contact
Gamma
1 11/16 in.
Not Autoclavable
3/8 in. ID tubing, 1/2 in. mini clamp
Fits 3/8 in. ID tubing
31/32 in.