Learn More
Cole-Parmer™ Sanitary Coupling with Barb, Polysulfone
Cole-Parmer™ Sanitary Coupling with Barb, Polysulfone has a tapered funnel design to provide a steady fluid flow.
1050.00 NOK - 1130.00 NOK
Specifications
Certifications/Compliance | Meets USP Class VI requirements and is FDA compliant for food contact |
---|---|
Autoclavable | Not Autoclavable |
Material | Polysulfone |
For Use With (Application) | FDA compliant for food contact |
Sterilization Method | Gamma |
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
12469650
|
Cole-Parmer™
31512-07 |
Fits 3/8 in. ID tubing |
1050.00 NOK
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||||
12479650
|
Cole-Parmer™
31512-08 |
Fits 1/2 in. ID tubing |
1130.00 NOK
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||||
Description
- Reduces dead space and improves flow.
- Tapered funnel design provides a steady fluid flow.
- Smooth inner surface prevents particle entrapment.
- Seamless construction prevents contamination by eliminating leaks and areas of potential bacterial growth.
- Material meets USP Class VI requirements and is FDA compliant for food contact.
Specifications
Meets USP Class VI requirements and is FDA compliant for food contact | |
Polysulfone | |
Gamma |
Not Autoclavable | |
FDA compliant for food contact | |
31/32 in. |