Learn More
Masterflex™ Sanitary Coupling
Unlike permanent couplers, these couplers can be attached to a new length of tubing when the old tubing wears out.
Brand:  Masterflex™ 31511-07
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12409690
  • 5260.00 NOK / Each
Estimated Shipment: 19-08-2024
Description
  • Suitable for clean-in-place, steam-in-place, and autoclaving for reuse; compatible with TRI-CLAMP™ fittings.
  • The internal stem is made of 316L stainless steel with an interior surface finish of 25RA or better. 
  • Fittings also feature an external sleeve made from a high-temperature, high-strength polymer and a PTFE O-ring to protect against any external entrapment.
  • Compression seal eliminates wicking between couplers and tubing.
  • Meets USDA compliance and 3A Sanitary Standards, and is FDA compliant
Specifications
Specifications
Vacuum Tubing
3/8 in.
5/8 in.
Sanitary
Sanitary Coupling, 3/8 in. ID, 5/8 in. OD, 3/8 in. Clamp
Stainless Steel
L/S 18, 35, 36 I/P 73 and B/T 86
3/8 in. Clamp