Learn More
Masterflex™ Sanitary Coupling
Unlike permanent couplers, these couplers can be attached to a new length of tubing when the old tubing wears out.
5260.00 NOK - 6675.00 NOK
Specifications
Product Type | Vacuum Tubing |
---|---|
Material | Stainless Steel |
Outlet Port | Sanitary |
Product Code | Brand | Diameter (English) Outer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) Outer | Price | Quantity & Availability | |||||
12409690
|
Masterflex™
31511-07 |
5/8 in. |
5260.00 NOK
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
12459690
|
Masterflex™
31511-20 |
1 3/8 in. |
6675.00 NOK
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
Description
- Suitable for clean-in-place, steam-in-place, and autoclaving for reuse; compatible with TRI-CLAMP™ fittings.
- The internal stem is made of 316L stainless steel with an interior surface finish of 25RA or better.
- Fittings also feature an external sleeve made from a high-temperature, high-strength polymer and a PTFE O-ring to protect against any external entrapment.
- Compression seal eliminates wicking between couplers and tubing.
- Meets USDA compliance and 3A Sanitary Standards, and is FDA compliant
Specifications
Vacuum Tubing | |
Stainless Steel | |
Sanitary |