Learn More
SymLine™ Safety Waste Caps
Shop All SCAT Europe Products
Click to view available options
Thread Size:
GL45
S55
S60/61
S90
Includes:
1 GL25 (m), Filling Lance, Connector for Exhaust Filter
1 NPT 3/8 in. (f), Connection for Exhaust Filter
2 PFA-fittings, Tube Connector
2 PFA-fittings, Tube Connector, Collector for HPLC Waste
3 PFA-fittings, Collector for HPLC Waste
5 PFA-fittings, Collector for HPLC Waste
Level Control w/ Shut-off, Connector for Exhaust Filter
Level Control, Cable Connection, Connector for Exhaust Filter
Level Control, Connector for Exhaust Filter
Level Control, Filling Lance, Connector for Exhaust Filter
Unit Size:
1 sheet
Each
Description
- Available with different thread sizes for attachment to pipe system
- Different numbers of hose connections for disposal of liquid waste
- Available with electronic level sensors that indicate the degree of filling of the disposal container
- By using a 4-in-1 collector (306509), up to 4 Safety Waste Caps can be connected simultaneously to only one connection point on the pipe system
Specifications
Specifications
| For Use With (Equipment) | SymLine System for Capillaries with (OD) 1.6/2.3/3.2 mm |
| Includes | 2 PFA-fittings, Tube Connector, Collector for HPLC Waste |
| Thread Size | GL45 |
| For Use With (Application) | Waste Disposal |
| Type | Safety Waste Cap |