Learn More
SCAT Safety Waste Cap 'LISA', V3.0 Extension GL25 Connection
Shop All SCAT Europe Products
Click to view available options
Thread Size:
GL25
Unit Size:
Each
Description
- To be screwed in over the top connection of a conductive LISA
- Adapter between conductive LISA and modules of the SymLine disposal system
- Made of conductive PE-HD-EC material to prevent static discharge
LISA is the first modular system for HPLC waste! Occupational safety and environmental protection with the proven SCAT technology, with maximum flexibility: Simply add connections and components as required. So you collect solvents, acids and alkalis safely - at any time.
Specifications
Specifications
| For Use With (Equipment) | SCAT Safety Waste Cap 'LISA', V3.0 |
| Material | PTFE |
| Thread Size | GL25 |
| For Use With (Application) | HPLC Systems |
| Type | Thread Adapter |