Learn More
SCAT™ Safety Waste Cap 'LISA', V3.0 Electronic Level Control Extension
Shop All SCAT Europe Products
Click to view available options
Type:
Electronic Control
Level Control
Material:
PTFE
PTFE, Conductive
Unit Size:
Each
Pack of 2
Pack of 3
Description
LISA is the first modular system for HPLC waste, providing occupational safety and environmental protection with proven SCAT™ technology and maximum flexibility. Simply add connections and components as required so you can collect solvents, acids and alkalis safely at any time.
- Simply unscrew the blind plug of the LISA, insert the level control, and connect cable to the signal box
- Permanently displays the fill level in your disposal container and alerts when reached as soon as it is full
- Available in electrically conductive and non-electrically conductive versions
Specifications
Specifications
| For Use With (Equipment) | SCAT™ Safety Waste Cap 'LISA', V3.0 |
| Material | PTFE, Conductive |
| For Use With (Application) | HPLC Systems |
| Type | Level Control |