Learn More
SCAT HPLC Safety Caps
GREENER_CHOICE
Product Code. 10541728 Shop All SCAT Europe Products
Change view
Click to view available options
Includes:
1 PFA Fitting (3.2 mm OD), Air Valve
2 PFA Fittings (3.2 mm OD), Air Valve
3 PFA Fittings (1.6/2.3/3.2 mm OD), 2 PFA Blind Plugs, Air Valve
3 PFA Fittings (3.2 mm OD), Air Valve
4 PFA Fittings (3.2 mm OD), Air Valve
Unit Size:
Each
Product Code. Includes unitSize
10541728 2 PFA Fittings (3.2 mm OD), Air Valve Each
10851421 1 PFA Fitting (3.2 mm OD), Air Valve Each
10348149 2 PFA Fittings (3.2 mm OD), Air Valve Each
16554170 3 PFA Fittings (3.2 mm OD), Air Valve Each
15168299 3 PFA Fittings (1.6/2.3/3.2 mm OD), 2 PFA Blind Plugs, Air Valve Each
10839170 3 PFA Fittings (3.2 mm OD), Air Valve Each
10222440 4 PFA Fittings (3.2 mm OD), Air Valve Each
7 options
Product Code. 10541728
Brand: SCAT 107031
Only null left
Add to basket
Add to basket
These safety caps provide maximum safety for your HPLC solvent supply. Extract your solvents safely from bottles and reservoirs without being exposed to hazardous gases and solvent vapors. Complete with integrated air valve for pressure equalization.
  • High quality materials ensure maximum safety for lab and staff
  • 360° rotatable and up to 200°C autoclavable screw cap for maximum flexibility
  • Stable construction for highest safety and easy handling
  • Chemically resistant against aggressive organic solvents
  • Attractive design and clear arrangement of the connections
  • Improved design for easy connection of capillaries in various colors
  • Free air valve with change indicator included with every SafetyCap
Specifications
For Use With (Equipment) Lab Bottles
Includes 2 PFA Fittings (3.2 mm OD), Air Valve
Material PTFE
Thread Size (Metric) GLS80
For Use With (Application) HPLC Systems
Type Safety Cap II