Learn More
Deutsch Neumann™ Rubber Spatulas
Product Code. 10282191
Change view
Click to view available options
:
Dimensions (L x W): 7 x 38mm
Product Code.
10282191 Dimensions (L x W): 7 x 38mm
1 options
Product Code. 10282191
Brand: Deutsch Neumann™ 2260001
Only null left
Add to basket
Add to basket
Excellent for the cleaning of test tubes. Deutsch Neumann Rubber Spatulas are manufactured from grey natural rubber.
  • 5mm hole for insert
  • Durometer 40 Shore A
Specifications
Material Rubber
Product Type Test Tube Cleaner
Length (Metric) 7 mm