Learn More
Fisherbrand™ Rubber Stoppers With a Bore
Product Code. 11537612 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric) Outer:
6 mm
13 mm
35 mm
37 mm
41 mm
45 mm
53 mm
Unit Size:
Pack of 10
Pack of 2
Pack of 20
Pack of 5
Pack of 50
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Unit Size
11537612 41 mm Pack of 5
11597592 13 mm Pack of 20
10283751 35 mm Pack of 10
11517612 37 mm Pack of 10
10000331 37 mm Pack of 10
11557612 45 mm Pack of 5
10080361 53 mm Pack of 2
10440123 6 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11537612
  • ONLINE EXCLUSIVE PRICE
    244.65 NOK
    194.34 NOK / Pack of 5
    Save 50.31 NOK
    20% Off
View all promotions
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
Prevents liquid chemical leaks or escape from container.
  • Used for industrial purposes
  • Fits tightly to the container
  • Red natural rubber construction
Specifications
Quantity 5/Pk.
Color Red
Diameter (Metric) Outer 41 mm
Material Rubber
Product Type Stopper (1 hole)