Learn More
Fisherbrand™ Rubber Stoppers With a Bore
Shop All Fisherbrand Products
Click to view available options
Diameter (Metric) Outer:
6 mm
13 mm
35 mm
37 mm
41 mm
45 mm
53 mm
Unit Size:
Pack of 10
Pack of 2
Pack of 20
Pack of 5
Pack of 50
Specifications
Specifications
| Quantity | 50/Pk. |
| Color | Red |
| Diameter (Metric) Outer | 6 mm |
| Material | Rubber |
| Product Type | Stopper (1 hole) |