Learn More
Fisherbrand™ Cork Stoppers
Product Code. 12366457 Shop All Fisherbrand Products
Click to view available options
Diameter (Metric) Bottom:
6 mm
8 mm
9 mm
12 mm
13 mm
14 mm
17 mm
19 mm
21 mm
22 mm
25.5 mm
28.5 mm
30 mm
32 mm
35 mm
38 mm
41.5 mm
44 mm
47.5 mm
50 mm
Diameter (Metric) Top:
9.5 mm
11 mm
12 mm
15 mm
16 mm
17 mm
20.5 mm
22 mm
24 mm
25.5 mm
28.5 mm
32 mm
33 mm
35 mm
38 mm
41.5 mm
44 mm
47.5 mm
50 mm
57 mm
Unit Size:
Pack of 10
Pack of 20
Product Code. 12366457
Only null left
Add to basket
Add to basket
For use in education, homebrewing and arts and crafts industries.
Available in different sizes
Specifications
Length (Metric) 22 mm
Quantity 20/Pk.
Diameter (Metric) Bottom 14 mm
Diameter (Metric) Top 17 mm
Material Cork
Product Type Cork Stopper