Learn More
Fisherbrand™ Cork Stoppers
Product Code. 12326467 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric) Bottom:
6 mm
8 mm
9 mm
12 mm
13 mm
14 mm
17 mm
19 mm
21 mm
22 mm
25.5 mm
28.5 mm
30 mm
32 mm
35 mm
38 mm
41.5 mm
44 mm
47.5 mm
50 mm
Diameter (Metric) Top:
9.5 mm
11 mm
12 mm
15 mm
16 mm
17 mm
20.5 mm
22 mm
24 mm
25.5 mm
28.5 mm
32 mm
33 mm
35 mm
38 mm
41.5 mm
44 mm
47.5 mm
50 mm
57 mm
Unit Size:
Pack of 10
Pack of 20
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Bottom Diameter (Metric) Top Unit Size
12326467 22 mm 25.5 mm Pack of 10
12346457 12 mm 15 mm Pack of 20
12356457 13 mm 16 mm Pack of 20
12366457 14 mm 17 mm Pack of 20
12386457 17 mm 20.5 mm Pack of 10
12316467 21 mm 24 mm Pack of 10
12336467 25.5 mm 28.5 mm Pack of 10
12306467 19 mm 22 mm Pack of 10
12346467 28.5 mm 32 mm Pack of 10
12356467 30 mm 33 mm Pack of 10
12803834 30 mm 33 mm Pack of 10
12366467 32 mm 35 mm Pack of 10
12376467 35 mm 38 mm Pack of 10
12386467 38 mm 41.5 mm Pack of 10
12396467 41.5 mm 44 mm Pack of 10
12306477 44 mm 47.5 mm Pack of 10
12316477 47.5 mm 50 mm Pack of 10
12306457 6 mm 9.5 mm Pack of 20
12326477 50 mm 57 mm Pack of 10
12316457 8 mm 11 mm Pack of 20
12326457 9 mm 12 mm Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
Product Code. 12326467
  • ONLINE EXCLUSIVE PRICE
    49.35 NOK
    37.77 NOK / Pack of 10
    Save 11.58 NOK
    23% Off
Estimated Shipment: 20-11-2026
Add to basket
Add to basket
For use in education, homebrewing and arts and crafts industries.
Available in different sizes
Specifications
Length (Metric) 32 mm
Quantity 10/Pk.
Diameter (Metric) Bottom 22 mm
Diameter (Metric) Top 25.5 mm
Material Cork
Product Type Cork Stopper