Learn More
ROBU Rubber Gaskets, GUKO
Conical gasket for filter funnels.
Brand:  ROBU 21857
Additional Details : Weight : 0.22320kg
 View more versions of this product
Product Code. 16893036
  • 330.00 NOK / Pack of 5
Estimated Shipment: 29-05-2024
Description
  • Conical
  • For filter funnels
Specifications
Specifications
Filter Funnel
5/Pk.
77 mm
Natural Rubber
57 mm