Learn More
ROBU Rubber Gaskets, GUKO
Conical gasket for filter funnels.
Brand:  ROBU 21854
Additional Details : Weight : 0.45000kg
 View more versions of this product
Product Code. 16684725
  • 220.00 NOK / Pack of 5
Estimated Shipment: 17-06-2024
Description
  • Conical
  • For filter funnels
Specifications
Specifications
Filter Funnel
5/Pk.
40 mm
Natural Rubber
26 mm