Learn More
ROBU Rubber Gaskets, GUKO
Conical gasket for filter funnels.
Brand:  ROBU 21851
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 16674725
  • 142.00 NOK / Pack of 5
Estimated Shipment: 25-06-2024
Description
  • Conical
  • For filter funnels
Specifications
Specifications
Filter Funnel
5/Pk.
22 mm
Natural Rubber
12 mm