Learn More
ROBU Rubber Gaskets, GUKO
Conical gasket for filter funnels.
Brand: ROBU 21857
Additional Details : Weight : 0.22320kg
Description
- Conical
- For filter funnels
Specifications
Filter Funnel | |
5/Pk. | |
77 mm |
Natural Rubber | |
57 mm |