Learn More
Fisherbrand™ Rubber Cone Filter Crucible Adapters
Product Code. 12318855 Shop All Fisherbrand Products
Change view
Click to view available options
Cone Size:
27 mm I.D., 18 mm I.D.
34 mm I.D., 23 mm I.D.
40 mm I.D., 25 mm I.D.
40 mm I.D., 27 mm I.D.
50 mm I.D., 30 mm I.D.
60 mm I.D., 40 mm I.D.
Height (Metric):
25mm
30mm
35mm
45mm
50mm
55mm
Unit Size:
Pack of 10
Product Code. Cone Size Height (Metric) unitSize
12318855 40 mm I.D., 25 mm I.D. 45mm Pack of 10
12388845 27 mm I.D., 18 mm I.D. 25mm Pack of 10
12308855 34 mm I.D., 23 mm I.D. 30mm Pack of 10
12328855 40 mm I.D., 27 mm I.D. 35mm Pack of 10
12338855 50 mm I.D., 30 mm I.D. 50mm Pack of 10
12348855 60 mm I.D., 40 mm I.D. 55mm Pack of 10
6 options
Product Code. 12318855
Only null left
Add to basket
Add to basket
Fisherbrand™ Rubber Cone Filter Crucible Adapters facilitate safe and secure placement of filter crucibles into a funnel.
  • Available in a range of sizes
  • Provided in packs of 10
Specifications
Cone Size 40 mm I.D., 25 mm I.D.
For Use With (Equipment) Filter crucibles
Height (Metric) 45mm
Material Rubber
Product Type Cone
Quantity 10Pack