missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPGRIP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13214-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a photoreceptor protein that interacts with retinitis pigmentosa GTPase regulator protein and is a key component of cone and rod photoreceptor cells. Mutations in this gene lead to autosomal recessive congenital blindness.Specifications
| RPGRIP1 | |
| Polyclonal | |
| Unconjugated | |
| RPGRIP1 | |
| CORD9, LCA6, RGI1, RGRIP, RPGR interacting protein 1, RPGRIP, RPGRIP1, RPGRIP1d | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 57096 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96KN7 | |
| RPGRIP1 | |
| RPGRIP1 Fusion Protein Ag4089 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction