missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This gene encodes a photoreceptor protein that interacts with retinitis pigmentosa GTPase regulator protein and is a key component of cone and rod photoreceptor cells. Mutations in this gene lead to autosomal recessive congenital blindness.
Specifications
Specifications
| Antigen | RPGRIP1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.41 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | RPGRIP1 |
| Gene Accession No. | Q96KN7 |
| Gene Alias | CORD9, LCA6, RGI1, RGRIP, RPGR interacting protein 1, RPGRIP, RPGRIP1, RPGRIP1d |
| Gene Symbols | RPGRIP1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?