Description
- Precision-engineered for optimal sample separation.
- All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
- Accessories (adaptors and buckets) further customize your centrifuge setup.
- Aerosol-tight options available.
Specifications
Specifications
| Type | Rotor |
| Capacity | 4 x 1,000 mL |
| Max. RCF | 4,347 x g |
| Max. Speed | 4,500 rpm |
| For Use With (Equipment) | Centrifuges 5910 Ri and 5920 R |
| Includes | Without Buckets |
| No. of Places | 4 |
| Model | S-4xUniversal |